Order Entry
Austria
ContactUsLinkComponent
Dulbecco's Modification of Eagle's Medium (1X), MP Biomedicals
Dulbecco's Modification of Eagle's Medium (1X), MP Biomedicals
Catalog # ICNA091642954
Supplier:  MP Biomedicals
CAS Number:  
undefined
Dulbecco's Modification of Eagle's Medium (1X), MP Biomedicals
Catalog # ICNA091642954
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    Dulbecco's Modified Eagle medium with 4,5 g/L dextrose, without L-glutamine and inositol
  • Storage temperature:
    4 °C

Specifications

About this item

Dulbecco's Modification of Eagle's Medium 1X with 4.5 g/L glucose, without L-glutamine, i-inositol is ideally suited for supporting and maintaining a vast range of mammalian cell types.

  • Most widely used modification of Eagle's Basal Medium (BME)
  • Contains four times greater concentration of amino acids, vitamins and supplementary components

The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.