ESD Safety Boots, Strap Closure, Mid, S3
: ELTEN
ESD safety boots feature a Velcro® closure. Available in white finish.
- Hydrophobised microfibre
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO grey
- MONO-PU sole SAFETY-GRIP
- Steel toe cap
Safety boots feature a non-metallic, penetration-resistant midsole.
: EN ISO 20345 S3 SRC, form B.
- Catalog No:
- ELTN760171/39
- ELTN760171/45
- ELTN760171/46
- ELTN760171/47
- ELTN760171/48
- ELTN760171/40
- ELTN760171/41
- ELTN760171/42
- ELTN760171/43
- ELTN760171/44
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together