Order Entry
Austria
ContactUsLinkComponent
QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit
QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit
Catalog # ABCAAB285293-96
Supplier:  Abcam
CAS Number:  
undefined
QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit
Catalog # ABCAAB285293-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA kit, Size; 100 assays

Specifications

About this item

A rapid sandwich ELISA kit to quantify CD8 in human serum, plasma, cell culture supernatnat.

  • Easy, convenient and time-saving method to measure the level of 24-OHC in human serum, plasma, culture supernatants and other biological fluids
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results
  • Detection Range: 0,5 to 100 ng/ml

An important cholesterol metabolite (cholesterol 24-hydroxylase) found in the brain. It is an endogenous agonist of the nuclear receptor LXR. It has been shown to induce cell death in neuroblastoma cells, possibly via a necroptosis pathway. 24(S)-Hydroxycholesterol has also been found to be a high affinity ligand for RORα and RORγ (Ki = 25 nM). 24(S)-Hydroxycholesterol levels are elevated in the cerebral spinal fluids of patients with neurogenerative diseases suggesting possible clinical applications. It was also able to reduce Aβ production and increase ER-resident immature amyloid precursor protein (APP) levels in human neuroblastoma SH-Sy5Y cells and CHO cells via inhibition of intracellular amyloid precursor protein trafficking. BioVision’s QuickDetect™ 24-OHC ELISA kit is a sandwich ELISA assay for measuring the quantity of human 24-OHC in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.