Order Entry
Austria
ContactUsLinkComponent
Cathepsin G (CTSG) (Human) ELISA Kit
Cathepsin G (CTSG) (Human) ELISA Kit
Catalog # ABCAAB285277-96
Supplier:  Abcam
CAS Number:  
undefined
Cathepsin G (CTSG) (Human) ELISA Kit
Catalog # ABCAAB285277-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Cathepsin G ELISA kit

Specifications

About this item

A sandwich ELISA kit for the quantitative measurement of cathepsin G in biological fluids.

  • Easy, convenient and time-saving method
  • Detection Range: 0,156 to 10 ng/ml
  • Sensitivity: <0,094 pg/ml
  • Assay precision: Intra-assay: CV <8%; inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed
  • 100 Assays per kit

Cathepsin G (CTSG) is an enzymatic protein belonging to the peptidase or protease families. The protein is found in azurophil granules of neutrophilic polymorphonuclear leukocytes.

The encoded protease has a specificity similar to that of Chymotrypsin C, and may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation.

CTSG ELISA kit is a sandwich ELISA assay for the quantitative measurement of human CTSG in serum, plasma, and cell culture supernatants.

The density of color is proportional to the amount of human CTSG captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.