Order Entry
Austria
Orders LinkContactUsLinkComponent
Phlebotomy/Sample Collection Tray

Phlebotomy/Sample Collection Tray

Phlebotomy/sample collection tray, 17 mm rack, 60 wells, handle height 241 mm, white, 419×311×85 mm
Catalog # HEAT120261
Supplier:   HEATHROW
Phlebotomy/Sample Collection Tray
Catalog # HEAT120261
Supplier:  HEATHROW
Phlebotomy/Sample Collection Tray
Phlebotomy/Sample Collection Tray

Phlebotomy/sample collection tray, 17 mm rack, 60 wells, handle height 241 mm, white, 419×311×85 mm

Price
211,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Organise your phlebotomy, I.V. start or catheter insertion supplies with this spacious, robust and easy to clean carry tray.

  • Lightweight easy-to-clean ABS construction
  • Comfort grip on durable acetal plastic handle tested to hold up to 10 kg
  • Comes with a 17 mm well, 60-place, tube rack

Customise the compartments to suit your requirements with the dividers provided.

Specifications
  • Description:
    Phlebotomy/sample collection tray, 17 mm rack, 60 wells, handle height 241 mm, white
  • W×D×H:
    419×311×85 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.