PicoKine Human APOE/Apolipoprotein E; ELISA Kit
Catalog # BSBTEK1455
Supplier: Boster Bio
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Human APOE/Apolipoprotein E PicoKine; ELISA kit
- Tests per kit:96 wells
Specifications
About this item
Sandwich high sensitivity ELISA kit for quantitative detection of human APOE.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect Human Apolipoprotein E/APOE with <50 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(heparin, EDTA), saliva, milk and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Apolipoprotein E/APOE tissue specificity: Occurs in all lipoprotein fractions in plasma. It constitutes 10-20% of very low density lipoproteins (VLDL) and 1-2% of high density lipoproteins (HDL). APOE is produced in most organs. Significant quantities are produced in liver, brain, spleen, lung, adrenal, ovary, kidney and muscle.
96wells/kit, with removable strips.