Order Entry
Austria
ContactUsLinkComponent
GFP PicoKine; ELISA Kit
GFP PicoKine; ELISA Kit
Catalog # BSBTEK1440
Supplier:  Boster Bio
CAS Number:  
undefined
GFP PicoKine; ELISA Kit
Catalog # BSBTEK1440
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Description:
    GFP PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of GFP.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect GFP GFP with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell lysates. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. GFP tissue specificity: Photocytes.

96wells/kit, with removable strips.