Order Entry
Austria
ContactUsLinkComponent
 
PicoKine Human Caspase 3; ELISA Kit
Catalog # BSBTEK1425
Supplier:  Boster Bio
CAS Number:  
undefined
PicoKine Human Caspase 3; ELISA Kit
Catalog # BSBTEK1425
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    Caspase-3
  • Description:
    Human Caspase 3 PicoKine; ELISA kit
  • Tests per kit:
    96 wells

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of human Caspase 3.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect Human Caspase 3/CASP3 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernate sand serum. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Caspase 3/CASP3 tissue specificity: Highly expressed in lung, spleen, heart, liver and kidney. Moderate levels in brain and skeletal muscle, and low in testis. Also found in many cell lines, highest expression in cells of the immune system.

96wells/kit, with removable strips.