Order Entry
Austria
ContactUsLinkComponent
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] solution staining reagent, for histology
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] solution staining reagent, for histology
Catalog # 13-0850-U500
Supplier:  Tonbo Biosciences
CAS Number:  
undefined
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] solution staining reagent, for histology
Catalog # 13-0850-U500
Supplier:  Tonbo Biosciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :557.467 g/mol
Storage Temperature:Freezer
CAS Number:150347-59-4
 :MFCD31812843
Formula:C₂₉H₁₉NO₁₁
MDL Number:MFCD00037466