Order Entry
Austria
ContactUsLinkComponent
NOS2/iNOS (Human) ELISA Kit
NOS2/iNOS (Human) ELISA Kit
Catalog # ABCAAB285315-96
Supplier:  Abcam
CAS Number:  
undefined
NOS2/iNOS (Human) ELISA Kit
Catalog # ABCAAB285315-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    NOS2/iNOS ELISA kit
  • Tests per kit:
    96 assays

Specifications

About this item

A sandwich ELISA kit for the quantitative measurement of NOS2 in biological fluids.

  • Easy, convenient and time-saving method
  • Detection Range: 78,125 TO 5000 pg/ml
  • Sensitivity: <46,875 pg/ml
  • Assay precision: Intra-assay: CV <8%, inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed

Nitric oxide synthases (NOS) are a family of enzymes that catalyse the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis.

In presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. There are three isoforms of NOS: Endothelial (eNOS), neuronal (nNOS), and inducible (iNOS).

iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as a part of body’s defense mechanism. NOS2 ELISA kit is a sandwich ELISA assay for the quantitative measurement of NOS2 in Human serum, plasma and cell culture supernatants.

The density of colour is proportional to the amount of NOS2 captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.