Order Entry
Austria
ContactUsLinkComponent
Collagen Type I (COL1)(rat) ELISA Kit, Inc.
Collagen Type I (COL1)(rat) ELISA Kit, Inc.
Artikelnr. ABCAAB285314-96
Lieferant:  Abcam
CAS-Nummer:  
undefined
Collagen Type I (COL1)(rat) ELISA Kit, Inc.
Artikelnr. ABCAAB285314-96
Lieferant:  Abcam
CAS-Nummer:  

Spezifikationen

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Description:
    Collagen type I ELISA kit
  • Tests per kit:
    100 assays

Spezifikationen

Über diesen Artikel

A sandwich ELISA kit for the quantitative measurement of collagen type I in biological fluids.

  • Easy, convenient and time-saving method
  • Detection range: 1,563 to 100 ng/ml
  • Sensitivity: <0,938 ng/ml
  • Assay precision: Intra-assay: CV <8%; Inter-Assay: CV <10%
  • No significant cross-reactivity or interference was observed

Collagen, a major component of the extracellular matrix, is a fibrous protein that provides tensile strength to tissues giving them structural integrity. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage.

All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.

The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (types I, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms.

COL1 ELISA kit is a sandwich ELISA assay for the quantitative measurement of COL1 in rat serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of COL1 captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.