Order Entry
Austria
ContactUsLinkComponent
Alanine Aminotransferase 1 (GPT) (rat) ELISA Kit
Alanine Aminotransferase 1 (GPT) (rat) ELISA Kit
Catalog # ABCAAB285264-96
Supplier:  Abcam
CAS Number:  
undefined
Alanine Aminotransferase 1 (GPT) (rat) ELISA Kit
Catalog # ABCAAB285264-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Target protein:
    Alanine Aminotransferase 1 (GPT)
  • Description:
    Alanine Aminotransferase 1 (GPT) (rat) ELISA Kit
  • Tests per kit:
    100 assays

Specifications

About this item

A Sandwich ELISA kit to the quantify rat GPT in serum, plasma and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of GPT in rat serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0,78 - 50 ng/ml
  • Sensitivity: < 0,469 ng/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%

Alanine aminotransferase 1 mediates the control of a large number of enzymes, ion channels, aquaporins and other proteins through calcium-binding. Among the enzymes to be stimulated by the calmodulin-calcium complex are a number of protein kinases and phosphatases. Together with CCP110 and centrin, is involved in a genetic pathway that regulates the centrosome cycle and progression through cytokinesis. BioVision’s GPT ELISA kit is a sandwich ELISA assay for the quantitative measurement of rat GPT in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of rat GPT captured from the samples.