Order Entry
Austria
ContactUsLinkComponent
QuickDetect™ Secretory IgA (Mouse) ELISA Kit
QuickDetect™ Secretory IgA (Mouse) ELISA Kit
Catalog # ABCAAB285304-96
Supplier:  Abcam
CAS Number:  
undefined
QuickDetect™ Secretory IgA (Mouse) ELISA Kit
Catalog # ABCAAB285304-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IgA
  • Description:
    QuickDetect™ Secretory IgA (Mouse) ELISA Kit
  • Tests per kit:
    100 assays

Specifications

About this item

A rapid sandwich ELISA kit for quantifying of secretory IgA in biological fluid.

  • Easy, convenient and time-saving method
  • Detection Range: 30 - 2000 pg/ml
  • Sensitivity: < 6 pg/ml

Immunoglobulin A (IgA) has two subclasses (IgA1 and IgA2) and can be produced as a monomeric as well as a dimeric form. The IgA dimeric form is the most prevalent and is also called secretory IgA (sIgA). sIgA is the main immunoglobulin found in mucous secretions, including tears, saliva, sweat, colostrum and secretions from the genitourinary tract, gastrointestinal tract, prostate and respiratory epithelium. It is also found in small amounts in blood. The secretory component of sIgA protects the immunoglobulin from being degraded by proteolytic enzymes, thus sIgA can survive in the harsh gastrointestinal tract environment and provide protection against microbes that multiply in body secretions.

BioVision's sIgA ELISA kit is a sandwich ELISA assay for the quantitative measurement of sIgA in Mouse serum, plasma and cell culture supernatants in 90 minutes. The density of color is proportional to the amount of sIgA captured from the samples.

Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed.Assay Precision: Intra-Assay: CV < 10%; Inter-Assay: CV < 12% (CV (%) = SD/mean X 100).