- Pk:96 Tests
- Assay Type:Sandwich
- Conjugate ELISA:HRP-Streptavidin
- Primary antibody reactivity:Human
- Target protein:Oxidized LDL
- Description:OxLDL (Human) ELISA kit
- Storage conditions:4 °C
- Sample type:Human serum, plasma, culture supernatants & biological fluids
- Detection method:Absorbance (450 nm)
- Detection range:3,125 - 200 ng/ml
A Sandwich ELISA kit for the quantitative measurement of OxLDL in human serum, biological fluids.
- Easy, convenient and time-saving method to measure the level of OxLDL in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
- Detection Range: 3,125 to 200 ng/ml
- Sensitivity: < 1,875 ng/ml
- Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%
- This assay has high sensitivity and excellent specificity for detection of OxLDL. No significant cross-reactivity or interference between OxLDL and analogues was observed
Low-density lipoprotein (LDL) are one of the five major groups of lipoprotein which transport all fat molecules around the body in the extracellular water. LDL delivers fat molecules to the cells and can drive the progression of atherosclerosis if they become oxidized within the walls of arteries. Since the Oxidized low-density lipoproteins (OxLDL) are more easily retained by the proteoglycans, a risk for cardiovascular disease when they invade the endothelium. BioVision’s OxLDL ELISA kit is a sandwich ELISA assay for the quantitative measurement of OxLDL in human serum, plasma and cell culture supernatants. The density of color is proportional to the amount of OxLDL captured from the samples.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.