Order Entry
Austria
ContactUsLinkComponent
 
Cyclic AMP Select ELISA Kit
 
undefined
Cyclic AMP Select ELISA Kit

 

  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Target protein:
    Cyclic AMP (cAMP)
  • Detection method:
    Colorimetric at 405 - 420 nm
  • Developmental time:
    90 - 120 min
  • Detection range:
    0,09 - 200 pmol/ml
  • Storage temperature:
    –20 °C
  • Sensitivity:
    80% B/B0: 0,6 pmol/ml
  • Incubation time:
    8 hr

 

Frequently Bought Together