DNA/RNA Oxidative Damage (Clone 7E6.9) ELISA Kit
Supplier: Cayman Chemical
Certificates
About this item
For the measurement of 8-hydroxy-2'-deoxyguanosine and 8-hydroxyguanosine.
- Measure DNA oxidative damage marker 8-hydroxy-2’-deoxyguanosine and RNA damage marker 8-hydroxyguanosine with equal selectivity and sensitivity
- Cross reactivity of clone 7E6.9 validated by mass spectrometry
- Assay 24 samples (urine or other matrices) in triplicate or 36 samples in duplicate
- Measure 8-hydroxyguanosine levels down to 0,45 ng/ml
- Incubation: 18 hours | Development: 90 to 120 minutes | Read: Colorimetric at 405 to 420 nm
Cayman's DNA/RNA Oxidative Damage (Clone 7E6.9) ELISA Kit is a competitive assay that can be used to measure both 8-hydroxy-2'-deoxyguanosine and 8-hydroxyguanosine. This kit has been validated for use in urine, but other sample types may be used. The assay has a range from 0,15 to 20 ng/ml and a sensitivity (80% B/B0) of approximately 0,45 ng/ml.
Delivery: Kit contains: Antibody, tracer, standard, buffers, reagents, and plate(s).
Specifications
- Assay Type:Competitive
- Format:Pre-coated
- Target protein:8-Hydroxyguanosine (8-OHG)
- Detection method:Colorimetric at 405 - 420 nm
- Developmental time:90 - 120 min
- Detection range:0,15 - 20 ng/ml
- Storage temperature:–20 °C
- Sensitivity:80% B/B0: 0,45 ng/ml
- Incubation time:18 hr
Specifications
Frequently Bought Together