ContactUsLinkComponent
Imhoff Cone without Stopcock
Imhoff Cone without Stopcock
Catalog # HECH44275010
undefined
Imhoff Cone without Stopcock
Catalog # HECH44275010
Specifications
  • Type:
    Without stopcock
  • Description:
    Imhoff cone, scale up to 1000 ml, ring mark at 1000 ml
  • Pk:
    1
Specifications
About this item
Made from borosilicate 3.3 glass.
  • Very good chemical resistance
  • Excellent thermal resistance
  • With frosted label and scale
  • Fused white graduation
The sedimentation cone with graduation according to Imhoff is used for the quantification of sediments from a suspension.
Certifications: Produced according to DIN 12672.