PicoKine Human CXCL4/PF4; ELISA Kit
Artikelnr. BSBTEK0726
Lieferant: Boster Bio
CAS-Nummer:
Spezifikationen
- Pk:1 KIT
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Human CXCL4/PF4 PicoKine; ELISA kit
- Tests per kit:96 wells/kit
Spezifikationen
Über diesen Artikel
Sandwich high sensitivity ELISA kit for quantitative detection of human CXCL4/PF4.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect human Cxcl4/PF4 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cxcl4/PF4 tissue specificity:
96wells/kit, with removable strips.