Order Entry
Austria
ContactUsLinkComponent
QuickDetect™ beta-Endorphin (Human) ELISA Kit
QuickDetect™ beta-Endorphin (Human) ELISA Kit
Catalog # ABCAAB287794-96
Supplier:  Abcam
CAS Number:  
undefined
QuickDetect™ beta-Endorphin (Human) ELISA Kit
Catalog # ABCAAB287794-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    QuickDetect™ beta-Endorphin (Human) ELISA Kit, 100 assays

Specifications

About this item

A rapid ELISA kit to quantify of beta-Endorphin in human serum, plasma, urine, cell culture samples.

  • Easy, convenient and time-saving method to measure the level of beta-Endorphin in human Serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 3 to 200 pg/ml
  • Sensitivity: <0,5 pg/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed

β-Endorphin (beta-EP) is an endogenous opioid neuropeptide and peptide hormone that is produced in certain neurons within the central nervous system and peripheral nervous system. It is one of five endorphins that are produced in human, the others of which include α-endorphin, γ-endorphin, α-neoendorphin, and β-neoendorphin. Function of β-endorphin has been known to be associated with hunger, thrill, pain, maternal care, sexual behavior, and reward cognition. In the broadest sense, β-endorphin is primarily utilized in the body to reduce stress and maintain homeostasis. In behavioral research, studies have shown that β-endorphin is released via volume transmission into the ventricular system in response to a variety of stimuli, and novel stimuli in particular. BioVision’s beta-EP ELISA kit is a sandwich ELISA assay for the quantitative measurement of beta-EP in human serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of beta-EP captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.