Order Entry
Austria
ContactUsLinkComponent
Ovalbumin (OVA) (Human) ELISA Kit
Ovalbumin (OVA) (Human) ELISA Kit
Catalog # ABCAAB289830-100
Supplier:  Abcam
CAS Number:  
undefined
Ovalbumin (OVA) (Human) ELISA Kit
Catalog # ABCAAB289830-100
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Ovalbumin (OVA) (Human) ELISA Kit, 100 assays

Specifications

About this item

A Sandwich ELISA kit for quantify Ovalbumin in human culture supernatants and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of Ovalbumin in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 0,781 to 50 ng/ml
  • Sensitivity: <0,469 ng/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • This assay has high sensitivity and excellent specificity for detection of OVA. No significant cross-reactivity or interference between OVA and analogues was observed

Ovalbumin (OVA) is the main protein found in egg white, making up approximately 55% of the total protein. Ovalbumin displays sequence and three-dimensional homology to the serpin superfamily, but unlike most serpins it is not a serine protease inhibitor. The function of ovalbumin is unknown, although it is presumed to be a storage protein. It is commonly used to stimulate an allergic reaction in test subjects as well as an established model allergen for airway hyper-responsiveness (AHR). BioVision’s Ovalbumin ELISA kit is a sandwich ELISA assay for the quantitative measurement of human Ovalbumin in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of human Ovalbumin captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.