ContactUsLinkComponent
Jenway® pH Electrodes
New Product
undefined
Jenway® pH Electrodes
New Product
  • Catalog No:
  • JENW99960-41
  • JENW99960-34
  • JENW90269-17
  • JENW68006-37
  • JENW68006-40
  • JENW59530-70
  • JENW99960-44
  • JENW10090-75
  • JENW99960-39
  • Model:
  • 99960-34
  • 59530-70
  • Connection:
  • BNC
  • BNC
  • BNC
  • 2 mm plug
  • S7
  • 3150/3205 waterproof connector
  • 3150/3205 waterproof connector
  • BNC/8 pin mini-DIN
  • BNC
  • Cable length:
  • 1 m
  • 1 m
  • pH range:
  • 0 - 14 pH
  • 0 - 14 pH
  • Temperature range:
  • 0...100 °C
  • Reference electrolyte:
  • AgCl, gel filled single junction
  • AgCl
Frequently Bought Together