
Trisodium naphthalene-1,3,5-trisulphonate ≥96%
Catalog # APOSOR18814-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:434.3056 g/mol
:GHS07
CAS Number:10533-44-5
Formula:C₁₀H₅O₉S₃·3Na
MDL Number:MFCD00065323