Order Entry
Austria
ContactUsLinkComponent
Pierce TMT™ Elution Buffer
Pierce TMT™ Elution Buffer
Catalog # PIER90104
CAS Number:  
undefined
Pierce TMT™ Elution Buffer
Catalog # PIER90104
CAS Number:  

Specifications

  • Pk:
    20 ml
  • Description:
    TMT elution buffer, for use with Anti-TMT™ resin
  • Seq:
    04

Specifications

About this item

Thermo Scientific TMT Elution Buffer is for use with Thermo Scientific Immobilized Anti-TMT Resin to enrich samples for peptides that have been chemically labeled with Tandem Mass Tag™ (TMT™) Reagents.

  • For use with a mass spectrometer
  • Designed to enrich chemically labeled samples for peptides
  • Helps provide higher specific enrichment compared to acidic or basic elution buffers

Immobilized Anti-TMT Resin and TMT Elution Buffer are effective for reducing sample complexity, improving dynamic range and studying cysteine modifications such as S-nitrosylation, oxidation and disulfide bonds of protein samples labeled with iodoTMT Reagents. This approach of selective thiol labeling and affinity enrichment is similar to isotope-coded affinity tags (ICAT™ Method) but allows for higher multiplex quantitation.

Caution: For Research Use Only. Not for use in diagnostic procedures.