ContactUsLinkComponent
Small RNA Kits
Small RNA Kits
Catalog # AGLSDNF-470-0275
Supplier:  AGILENT
Small RNA Kits
Catalog # AGLSDNF-470-0275
Supplier:  AGILENT
Specifications
  • Description:
    Small RNA kit
  • Environmentally Preferable:
  • Pk:
    1
  • Details:
    Fragment analyser system kit for the quantitative and qualitative analysis of small and microRNAs from 15 - 200 nt, includes reagents necessary for running 275 samples
Specifications
About this item
Analysis of small RNA quality control including microRNA quantification.
  • Broad concentration range - The small RNA kit has a concentration range of 25 to 2500 pg/µl
  • Short analysis time - Analysis time in as little as 22 minutes
  • Reliable sizing - Sizing range from 15 to 200 nt ensures accurate and precise sizing of small RNA samples
  • Low sample volume - Only 2 µl of sample is required, minimising sample loss to QC steps
  • Specific RNA analysis modes - ProSize data analysis software uses a specific small RNA analysis region function automatically calculating percent microRNA and quantification of microRNA and small RNA
  • Capillary array - Optimised for the 12 -capillary array
The small RNA kit focuses on microRNA and small RNA quality control. Analysis of small RNAs is essential for downstream applications such as small RNA NGS libraries. The small RNA ladder includes six fragments ranging from 15 to 200 nt, which provides accurate and precise quantification and sizing of small RNA and microRNA. Focusing on a narrow range of 200 nt magnifies the individual areas of microRNA (10 to 40 nt) and small RNA (40 to 200 nt) allowing for high separation resolution and detailed analysis of each region.
Caution: For research use only. Not for use in diagnostic procedures.