VWR®, Peristaltic Pump Head
181-0291
- Maximum Flow Rate:1,26 L/min
- Description:Peristaltic pump head, easy tube loading, electric drive
- Pk:1
The pump heads are rugged fabrications of polyamide, acetal and stainless steel with an integral pump head drive shaft with dual ball race bearings.
Maximum head: 20 m
: The pumps are supplied with a stainless steel mounting plate.