Order Entry
Austria
ContactUsLinkComponent
Expression vectors, green renilla Luc vector for luciferase assays, Pierce™
Expression vectors, green renilla Luc vector for luciferase assays, Pierce™
Catalog # PIER16152
CAS Number:  
undefined
Expression vectors, green renilla Luc vector for luciferase assays, Pierce™
Catalog # PIER16152
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Description:
    pMCS-Green Renilla Luc Vector
  • Seq:
    01
  • Storage temperature:
    −70...−20 ˚C

Specifications

About this item

Green Renilla Luc vectors contain a gene cloned from the sea pansy, Renilla reniformis

  • Intracellular Green Renilla luciferase gene, optimised for high expression in mammalian systems
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimise background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells respectively
  • High-copy pUC bacterial DNA replication origin

pMCS-Renilla Luc vector is a multiple cloning site plasmid to designed to accept a promoter sequence for study of gene regulation using the intracellular Renilla luciferase reporter.

pCMV-Green Renilla Luc vector is a derivative of pMCS-Green Renilla Luc. It contains the luciferase gene under the control of the CMV (Cytomegalovirus) promoter. Strong (CMV) constitutive promoters for co-transfection and normalization.

pTK-Green Renilla Luc vector is a derivative of pMCS-Green Renilla Luc. It contains the luciferase gene under the control of the Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter. Weak (TK) constitutive promoters for co-transfection and normalization.

The gene encodes a bioluminescent Green Renilla luciferase (36 kDa), which enables measurement of the reporter activity in media (for real-time assays) and in cell lysates.

Caution: For research use only. Not for use in diagnostic procedures.