Order Entry
Austria
ContactUsLinkComponent
Ammonium acetate 5mM in water, HiPerSolv CHROMANORM®, PFAS grade eluent, for LC-MS
Ammonium acetate 5mM in water, HiPerSolv CHROMANORM®, PFAS grade eluent, for LC-MS
Catalog # 92500.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium acetate 5mM in water, HiPerSolv CHROMANORM®, PFAS grade eluent, for LC-MS
Catalog # 92500.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Nominal concentration (V/V):
    4.75 - 5.25 mM
  • Ca (Calcium):
    max. 0.2 ppm
  • K (Potassium):
    max. 0.2 ppm
  • Mg (Magnesium):
    max. 0.2 ppm
  • Na (Sodium):
    max. 0.5 ppm
  • Suitability for PFAS analysis:
    passes test

Specification Test Results

About this item

CAS Number:631-61-8
 :
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
 :77.08 g/mol
Storage Temperature:Ambient
EINECS:231-791-2