Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 47% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Irritant
Corrosive
Potassium hydroxide 47% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Potassium hydroxide 47% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (acidimetric):
    46.0 - 48.0 %
  • Carbonate (as K2CO3):
    max. 1.0 %
  • Chloride (Cl):
    max. 0.004%
  • Phosphate (PO4):
    max. 0.0005%
  • Sulfate (SO4):
    max. 0.002%
  • Total nitrogen (N):
    max. 0.0003%
  • Heavy metals (as Pb):
    max. 0.0005%
  • Al (Aluminium):
    max. 0.001%
  • As (Arsenic):
    max. 0.0001%
  • Ca (Calcium):
    max. 0.0005%
  • Cu (Copper):
    max. 0.0005%
  • Fe (Iron):
    max. 0.0005%
  • Mg (Magnesium):
    max. 0.0005%
  • Ni (Nickel):
    max. 0.0005%
  • Pb (Lead):
    max. 0.0005%
  • Zn (Zinc):
    max. 0.0005%

Specification Test Results

Related Information

Frequently Bought Together