Order Entry
Austria
ContactUsLinkComponent
 
Sodium acetate, anhydrous 99.0-101.0%, EMPROVE® EXPERT ChP, USP, SAFC®
 
Sodium acetate, anhydrous 99.0-101.0%, EMPROVE® EXPERT ChP, USP, SAFC®

 

  • Assay (perchloric acid titration, calculated on dried substance):
    99.0-101.0 %
  • Identity:
    passes test
  • Appearance:
    colorless crystals or white crystalline powder
  • In water insoluble matter:
    ≤ 0.05 %
  • pH-value (3 %, water):
    7.5 - 9.2
  • Chloride (Cl):
    ≤ 0.01 %
  • Sulfate (SO4):
    ≤ 0.0050 %
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • Al (Aluminium):
    ≤ 0.00002 %
  • As (Arsenic):
    s 0.0002 %
  • Fe (Iron):
    ≤ 0.001 %
  • K (Potassium):
    ≤ 0.003 %
  • Calcium and Magnesium:
    passes test
  • Assay (Acetic acid):
    ≤ 0.4 %
  • Loss on drying (120 °C):
    ≤ 1.0 %
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Endotoxins:
    ≤ 2.5 IU/g
  • Total aerobic microbial count (TAMC):
    ≤100 CFU/g
  • Total combined yeasts and molds count (TYMC):
    ≤ 100 CFU/g

 

Frequently Bought Together