ContactUsLinkComponent
Performance Professional Freezers with Static Cooling and Plastic Inner Liner, FFFsg
Performance Professional Freezers with Static Cooling and Plastic Inner Liner, FFFsg
Supplier:  Liebherr
Certificates
undefined
Performance Professional Freezers with Static Cooling and Plastic Inner Liner, FFFsg
Supplier:  Liebherr
Specifications
  • Catalog No:
  • LIEBFFFSG4001
  • LIEBFFFSG5501
  • LIEBFFFSG6501
  • Model:
  • FFFsg 4001
  • FFFsg 5501
  • FFFsg 6501
  • Refrigerant:
  • R 290
  • R 290
  • R 290
  • Insulation:
  • 80/67 mm
  • 63/67 mm
  • 63/67 mm
  • Display:
  • Monochrome display, touch buttons
  • Monochrome external digital display, touch buttons
  • Monochrome external digital display, touch buttons
  • Defrost:
  • Manual
  • Manual
  • Manual
  • Alarm:
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Electrical:
  • ~220 - 240 V, 50 Hz, 2,0 A
  • ~220 - 240 V, 50 Hz, 2,0 A
  • ~220 - 240 V, 50 Hz, 2,0 A
  • Shelves/drawers:
  • Adjustable, wire shelves plastic-coated
  • Adjustable, wire shelves plastic-coated, 10 baskets
  • Adjustable, wire shelves plastic-coated, 12 baskets
  • Door type:
  • Steel, self closing
  • Steel, self closing
  • Steel, self closing
  • Door lock:
  • Mechanical
  • Mechanical
  • Mechanical
  • Weight:
  • 80 (gross)/75 (net) kg
  • 99 (gross)/72 (net) kg
  • 119 (gross)/109 (net) kg