ContactUsLinkComponent
Spark-free Performance laboratory refrigerators with ATEX certificate, SRFfg
 :  Liebherr
undefined
Spark-free Performance laboratory refrigerators with ATEX certificate, SRFfg
 :  Liebherr
  • Catalog No:
  • 471-1340
  • LIEBSRFFG5501
  • LIEBSRFFG4001
  • Model:
  • SRFfg 3501
  • SRFfg 5501
  • SRFfg 4001
  • Display:
  • Monochrome display, touch buttons
  • Monochrome display, touch buttons
  • Monochrome display, touch buttons
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Alarm:
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Access ports:
  • 1ר 10 mm
  • 1ר 10 mm
  • 1ר 10 mm
  • Shelves/drawers:
  • 4 adjustable, glass shelves
  • 5 adjustable, glass shelves
  • 5 adjustable, glass shelves
  • Door type:
  • steel
  • steel
  • steel
  • Door lock:
  • mechanical
  • mechanical
  • mechanical
  • Refrigerant:
  • R 600a
  • R 600a
  • R 600a
  • Sound level:
  • 48 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • Electrical:
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • Weight:
  • 70 (gross)/60 (net) kg
  • 92 (gross)/80 (net) kg
  • 74 (gross)/68 (net)kg
Frequently Bought Together