ContactUsLinkComponent
Value Plastics® Adapter Fittings, Luer to Plug
Value Plastics® Fitting, Polypropylene, Straight, Male Luer Slip to Plug with Tether Loop Adapter; 1000/PK
Catalog # MFLX45518-70BLK
undefined
Value Plastics® Fitting, Polypropylene, Straight, Male Luer Slip to Plug with Tether Loop Adapter; 1000/PK
Catalog # MFLX45518-70BLK
Specifications
  • Connection Type:
    Male Luer Plug
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Male Luer Slip to Plug with Tether Loop Adapter; 1000/PK
  • Seq:
    02
  • Connection type (port 1):
    Male luer slip with tether loop
  • No. of ports:
    1
  • Pk:
    1.000
  • Fitting material:
    Polypropylene
Specifications
About this item
Full material traceability to ensure process integrity.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Connection Type: Male Luer to Plug with Tether Loop
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Connection Type:
    Male Luer Plug
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Male Luer Slip to Plug with Tether Loop Adapter; 1000/PK
  • Seq:
    02
  • Environmentally Preferable:
  • Connection type (port 1):
    Male luer slip with tether loop
  • No. of ports:
    1
  • Pk:
    1.000
  • Fitting material:
    Polypropylene