ContactUsLinkComponent
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene
Value Plastics® Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
Catalog # MFLX45508-30BLK
undefined
Value Plastics® Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
Catalog # MFLX45508-30BLK
Specifications
  • Barb size/tubing int.Ø (port 1):
    1/16"
  • Connection Type:
    Female Luer to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
  • Seq:
    02
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Hose barb
  • No. of ports:
    2
  • Pk:
    1.000
  • Fitting material:
    Polypropylene
Specifications
About this item
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
  • Barb size/tubing int.Ø (port 1):
    1/16"
  • Connection Type:
    Female Luer to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
  • Seq:
    02
  • Environmentally Preferable:
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    Hose barb
  • No. of ports:
    2
  • Pk:
    1.000
  • Fitting material:
    Polypropylene