ContactUsLinkComponent
Dual-wire wrist strap parking station, MagSnap 360™
Dual-wire wrist strap parking station, MagSnap 360™
Catalog # 115-3046
Supplier:  DESCO
Dual-wire wrist strap parking station, MagSnap 360™
Catalog # 115-3046
Supplier:  DESCO
Specifications
  • Description:
    MagSnap 360™ Dual-wire wrist strap parking station
  • Pk:
    1
Specifications
About this item
This provides a storage terminal for the wrist cord when the workstation is unoccupied.
  • Mounts to any surface conveniently located nearby the operator
  • Spares operator jacks from the mechanical wear and tear of constant insertion and removal of the wrist cord
  • Monitors display a PASS condition for the operator when the wrist cord is parked
Gross weight: 0,01 kg.