ContactUsLinkComponent
Dual-wire thermoplastic wrist strap
Dual-wire thermoplastic wrist strap
Catalog # 115-3067
Supplier:  DESCO
undefined
Dual-wire thermoplastic wrist strap
Catalog # 115-3067
Supplier:  DESCO
Specifications
  • Colour:
    Blue
  • Description:
    Dual wire adjustable thermoplastic wrist strap
  • Pk:
    1
Specifications
About this item
The Dual-Wire thermoplastic wrist strap consists of an insulative exterior with a conductive interior and a 3,4 mm dual conductor jack. The wrist strap is lightweight, low profile, and safely makes contact with human skin.
  • Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
  • Zipper style latching mechanism easily adjusts to most wrist sizes
Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of EN 61340-5-1 and Wrist Strap standard IEC 61340-4-6.
Ordering information: Designed for use with SCS Dual-Wire Coil Cords (sold separately).