Dual-wire fabric wristband, SCS
: DESCO
The Dual-wire fabric wristband is available in one size that adjusts to almost any size wrist for reliable protection.
- Adjustable, elastic nylon material with conductive woven thread
- Fits most wrist sizes to provide reliable protection
- Dual-Wire conductor provides redundancy
- Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
The wristband provides full conductivity, comfort, and reliability, while ensuring continuous drain of static charge when used with SCS 2300 Dual-wire coil cords (sold separately). Made to be used in conjunction with all SCS workstation monitors.
Wristband contact 360 degrees around wrist, plug size of 3,4 mm and cord removal force of 1 to 5 pounds.
: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.
Frequently Bought Together