ContactUsLinkComponent
Single wire disposable wrist strap, SCS
Single wire disposable wrist strap, SCS
Catalog # 115-3055
Supplier:  DESCO
undefined
Single wire disposable wrist strap, SCS
Catalog # 115-3055
Supplier:  DESCO
Specifications
  • Description:
    Single wire, disposable wrist strap, 1,52 m
  • Pk:
    1
Specifications
About this item
Wrist strap (2209) is designed for one-time use. The wrist strap is ideally suited for shipment along with ESD sensitive items [ESDS], providing the means to ground personnel opening ESDS packaging and installing contents.
  • Disposable non-woven fabric wrist strap provides cost-effective
  • Hypo-allergenic adhesive provides 360 degree contact with skin
  • Minimizes skin-to-band resistance
  • Conductive adhesive on the copper foil adheres to any convenient electrical ground
Use for visitors and one-time use applications. Suited for shipment along with components and devices, such as memory upgrades, sound cards, etc., to provide protection at the receiving end. Equipment chassis to provide equipotential bonding. A current limiting resistance is fabricated into the plastic ribbon below the wristband.
Certifications: Meets the ANSI/ESD S20.20 wrist strap system required limit tested per ESD TR53. UL listed.
Packaging: Individually packaged.