Silicone encased keyboards with full layout, integrated touch pad and backlit keys, GCQ CleanType® Prime Touch+
Supplier: GETT GERATETECHNIK
Certificates
About this item
Silicone encased keyboard with full layout, integrated touch pad and backlit keys for use in highly hygienic sensitive areas. Completely waterproof and dust proof.
- Integrated capacitive touch pad
- Five-stage lighting function
- Flexible mounting using magnets
- EMC according to EN 60601-1-2 Edition 4
- Keyboard meets IP 68 protection rating due to a fully enclosed silicone surface
- Fully washable and disinfectable
- Cleaning possible without unintentionally triggering keys by activating a keylock button
- Two-year warranty
- Engineered in Germany
The Cleantype® Prime Touch+ keyboard helps create a hygienic working environment. The surface is waterproof and dust proof and can therefore be thoroughly cleaned and disinfected.
This model keyboard is also equipped with special product features. It can be attached to magnetic surfaces in a flexible way and is suitable for darkened environments where space is cramped thanks to an incremental lighting function and the slim design. Thanks to the touch pad, there is no need to use an additional mouse. It provides the writing comfort of an office keyboard.
Certifications: CE, EMC, REACH, IP 68.
Ordering information: Includes USB extension cable.
Packaging: Keyboards are packed in brown cardboard boxes.
Frequently Bought Together