Orders LinkContactUsLinkComponent
Stopcocks for dessicator lids
Stopcocks for dessicator lids
Supplier:   witeg Labortechnik
Stopcocks for dessicator lids
Supplier:  witeg Labortechnik
Certificates
Stopcocks for dessicator lids
Stopcocks for dessicator lids
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • For
    Availability
    Price
  • WITG0.246.002
    Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
    Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,20 €
    41,20 €
    1 * 1 ST
       41,20 €
  • 467-1255
    Stopcock for desiccator lids, borosilicate glass, with PTFE needle valve for thread connection with over twist
    Stopcock for desiccator lids, borosilicate glass, with PTFE needle valve for thread connection with over twist
    MOBILEX
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,90 €
    34,90 €
    1 * 1 ST
       34,90 €