Order Entry
Austria
Orders LinkContactUsLinkComponent
pH Cleaning Solution Set

pH Cleaning Solution Set

pH cleaning solution set (2×500 ml solution A+B + 1×500 ml storage solution), 1 SET
Catalog # HAMI238290
Supplier:   HAMILTON BONADUZ
pH Cleaning Solution Set
Catalog # HAMI238290
Supplier:  HAMILTON BONADUZ
CAS Number:  
pH Cleaning Solution Set
pH Cleaning Solution Set

pH cleaning solution set (2×500 ml solution A+B + 1×500 ml storage solution), 1 SET

CAS Number:  
Price
166,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

A two-part cleaning solution for all glass membrane pH sensors. pH Cleaning solution A contains sodium hydroxide (<3%), pH cleaning solution B contains hydrochloric acid (4%).

Also includes storage solution.

Delivery information: Includes 2 × 500 ml A+B cleaning solutions + 1 × 500 ml storage solution.

Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    pH cleaning solution set (2×500 ml solution A+B + 1×500 ml storage solution)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.