ContactUsLinkComponent
Cleaning and Storage Solutions
Cleaning and Storage Solutions
Catalog # 662-9223
Supplier:  HAMILTON BONADUZ
CAS Number:  
undefined
Cleaning and Storage Solutions
Catalog # 662-9223
Supplier:  HAMILTON BONADUZ
CAS Number:  
Specifications
  • Pk:
    500 ml
  • Description:
    Electrode storage solution, 500 ml, for pH and ORP sensors
  • Seq:
    22
Specifications
About this item
Hamilton storage solution is recommended to increase sensor longevity when not in use. The liquid solution is acid buffered to help with sensor response time after storage as well as prevent any biological growth.