Order Entry
Austria
ContactUsLinkComponent
L-Arginine hydrochloride, Multi-Compendial, J.T.Baker®
L-Arginine hydrochloride, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
L-Arginine hydrochloride, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes test
  • USP - Specific Rotation [Å]²º^D (+):
    21.4 - 23.6 °
  • USP - Loss on Drying at 105°C:
    ≤ 0.2 %
  • USP - Residue on Ignition:
    ≤ 0.1 %
  • USP - Sulfate (SO₄):
    ≤ 0.03 %
  • USP - Related Compounds - Individual Impurities:
    ≤ 0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤ 2.0 %
  • USP - Chloride (Cl):
    16.5 - 17.1 %
  • EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes test
  • EP/BP - Identification B:
    Passes test
  • EP/BP - Identification E:
    Passes test
  • EP/BP - Appearance of Solution:
    Passes test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    21.0 - 23.5 °
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤ 0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤ 0.5 %
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤ 0.02 %
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis):
    98.5 - 101.0 %
  • JP - Identification:
    Passes test
  • JP - Optical Rotation (+):
    21.5 - 23.5 °
  • JP - pH (1 in 10):
    4.7 - 6.2
  • JP - Clarity and Color of Solution:
    Passes test
  • JP - Sulfate (SO₄):
    ≤ 0.028 %
  • JP - Ammonium (NH₄):
    ≤ 0.02 %
  • JP - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Related Substances:
    Passes test
  • JP - Loss on Drying at 105°C:
    ≤ 0.20 %
  • JP - Residue after Ignition:
    ≤ 0.10 %
  • Endotoxin Concentration, IU/mg, :
    For Information Only

Specification Test Results

Related Information

Frequently Bought Together