Orders LinkContactUsLinkComponent
Bucket, Rectangular
Bucket, Rectangular
Supplier:   FHP VILEDA SCS
Bucket, Rectangular
Supplier:  FHP VILEDA SCS
Certificates
Bucket, Rectangular
Bucket, Rectangular
2 Options Available Below
Product Details & Documents
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE pre-prepared trolley.
  • Liquid measurement lines molded inside of bucket
  • Color coding clips included to allow differentiation in colours of buckets (red, blue, green, yellow)
  • Autoclavable
Highest performance for the best contamination control.
Product Family Options
Product Information
  • Colour
    Capacity
    Availability
    Price
  • 129-0458
    Bucket with colour coding clips
    Bucket with colour coding clips
    Grey
    6 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,00 €
    25,00 €
    1 * 1 ST
       25,00 €
  • 129-0454
    Long Bucket with colour coding clips
    Long Bucket with colour coding clips
    Grey
    22 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,50 €
    58,50 €
    1 * 1 ST
       58,50 €