ContactUsLinkComponent
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Supplier:  Tecan
Certificates
undefined
Spark® Cyto multimode plate readers with live cell imaging and real-time cytometry
Supplier:  Tecan
Specifications
  • Catalog No:
  • TCAT94400001
  • TCAT94300001
  • TCAT94200001
  • TCAT94100001
  • TCAT94000001
  • Model:
  • Spark® Cyto 600
  • Spark® Cyto 500
  • Spark® Cyto 400
  • Spark® Cyto 300
  • Spark® Cyto 100
  • Light source:
  • High energy xenon flash lamp
  • High energy xenon flash lamp
  • High energy xenon flash lamp
  • High energy xenon flash lamp
  • High energy xenon flash lamp
  • Measuring modes:
  • Luminescence, Fluorescence, Absorbance
  • Luminescence, Fluorescence, Absorbance
  • Luminescence, Fluorescence, Absorbance
  • Luminescence, Fluorescence, Absorbance
  • Luminescence, Fluorescence, Absorbance
  • Wavelength selection:
  • Monochromator and filter optics (fluorescence)
  • Filter optics (fluorescence)
  • Monochromator optics (fluorescence)
  • Filter optics (fluorescence)
  • Interfaces:
  • USB 3.0
  • USB 3.0
  • USB 3.0
  • USB 3.0
  • USB 3.0
  • Languages:
  • English, German, French, Italian, Spanish, Portuguese
  • English, German, French, Italian, Spanish, Portuguese
  • English, German, French, Italian, Spanish, Portuguese
  • English, German, French, Italian, Spanish, Portuguese
  • English, German, French, Italian, Spanish, Portuguese
  • Plate types:
  • Nano-Quant Plate
    From 1-well to 1536-well SBS plates
    Imaging from 6-well to 384-well SBS plates
  • Nano-Quant Plate
    From 1-well to 1536-well SBS plates
    Imaging from 6-well to 384-well SBS plates
  • Nano-Quant Plate
    From 1-well to 1536-well SBS plates
    Imaging from 6-well to 384-well SBS plates
  • Nano-Quant Plate
    From 1-well to 384-well SBS plates
    Imaging from 6-well to 384-well SBS plates
  • Nano-Quant Plate
    Imaging from 6-well to 384-well SBS plates
  • Shaking:
  • Linear, orbital, and double orbital
  • Linear, orbital, and double orbital
  • Linear, orbital, and double orbital
  • Linear, orbital, and double orbital
  • Linear, orbital, and double orbital