Order Entry
Austria
ContactUsLinkComponent
Microplate reader, Infinite® F50 Plus
Microplate reader, Infinite® F50 Plus
Catalog # TCAT91000001
Supplier:  Tecan
undefined
Microplate reader, Infinite® F50 Plus
Catalog # TCAT91000001
Supplier:  Tecan

Specifications

  • Model:
    Infinite F50 Plus
  • Light source:
    LED, self-calibrating
  • Plate types:
    96-well plates
  • Wavelength selection:
    8-channel optics + reference channel
  • Description:
    Singlemode microplate reader for measuring of absorbance with data analysis software Magellan V7.5 STD 2 PC and temperature control
  • Seq:
    01
  • Bandwidth:
    10 ±2 nm
  • Filters:
    Standard filter: 405, 450, 492, 620 nm
  • Interfaces:
    USB 2.0
  • Shaking:
    Linear plate shaking in four different modes
  • Temperature operating range:
    15...35 °C
  • Wavelength range:
    400 - 750 nm
  • Weight:
    2,6 kg
  • W×D×H:
    347×189×134 mm
  • Pk:
    1

Specifications

About this item

The Tecan Infinite F50 Plus absorbance plate reader, together with the Magellan data analysis software, is an ideal solution for photometric measurements of ELISAs and kinetic assays in the 400 to 750 nm wavelength range.

  • Ideal for absorbance-based ELISAs, kinetic and multi-label reading
  • 405, 450, 492 and 620 nm filters included
  • Ready for qualitative and quantitative data analysis using Magellan data analysis software
  • 8-channel optics + reference channel for fast reading & optimized light intensity
  • Linear plate shaking
  • Designed to meet IVDR (EU) 2017/746

The Infinite F50 Plus is a reliable, validated and compact absorbance microplate reader for ELISA. The reader provides accurate, fast photometric measurements using innovative eight-channel optics and LED technology.

Together with Magellan™ reader control and data analysis software, this reader is ideal for a variety of ELISA applications. The compact Infinite F50 Plus fits into any lab, saving valuable bench space.

Two year warranty and tested reliability of >100,000 plates.

Ordering information: Accessories not included (must be ordered seperately). Additional configuration available on request.
Delivery: Includes 1× reader, 1× Magellan data analysis software, 1× power cable, 1× USB stick with manuals in 11 languages.