Potassium chloride, Sigma-Aldrich®
SIALBCR113-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:74.55 g/mol
:14,07621
:1420 °C (1013 hPa)
:773 °C
:1.98 g/cm³ (20 °C)
:Ambient
:231-211-8
:7447-40-7
:KCl
:MFCD00011360