Order Entry
Austria
ContactUsLinkComponent
VWR®, ICP-OES Wavelength Calibration Solution Concentrate, 15 components in 5% HNO₃
VWR®, ICP-OES Wavelength Calibration Solution Concentrate, 15 components in 5% HNO₃
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, ICP-OES Wavelength Calibration Solution Concentrate, 15 components in 5% HNO₃
Supplier:  VWR Chemicals
Certificates

Specifications

  • Hazard class description:
    Corrosive
  • UN No.:
    3264
  • Storage temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together