VWR®, ICP Calibration Standard - Quality Control (XVI), 21 Elements: 100 mg/l each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in 2% HNO₃
Catalog # 91266.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Matrix:2% HNO₃
- Pack type:HDPE bottle
- Pk:100 mL
- Description:ICP Calibration Standard - Quality Control (XVI) (21 Elements)
- Seq:02
- Hazard class description:Corrosive
- UN No.:3264
- Storage temperature:Ambient
- Elements and Concentration:100 mg/l each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn
Specifications
About this item
Certifications: ISO 17025/17034