Order Entry
Austria
ContactUsLinkComponent
VWR®, Instrument Calibration Standard 2, 26 Elements in 5% HNO₃/tr. HF/tr. C₄H₆O₆
VWR®, Instrument Calibration Standard 2, 26 Elements in 5% HNO₃/tr. HF/tr. C₄H₆O₆
Catalog # 91198.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Instrument Calibration Standard 2, 26 Elements in 5% HNO₃/tr. HF/tr. C₄H₆O₆
Catalog # 91198.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr. HF/tr. C₄H₆O₆
  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 2 (26 Elements)
  • Seq:
    02
  • Hazard class description:
    Corrosive
  • UN No.:
    3264
  • Storage temperature:
    Ambient
  • Elements and Concentration:
    100 ug/ml each of Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn

Specifications

About this item

26 components: 100 ug/ml each of Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn in 5% HNO₃/tr. HF/tr. C₄H₆O₆

Certifications: ISO 17025/17034