Order Entry
Austria
ContactUsLinkComponent
VWR®, NexION Setup Solution, 8 Elements: 1 ug/l each of Be, Ce, Fe, in, Li, Mg, Pb, U in 1% HNO₃
VWR®, NexION Setup Solution, 8 Elements: 1 ug/l each of Be, Ce, Fe, in, Li, Mg, Pb, U in 1% HNO₃
Catalog # 91181.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, NexION Setup Solution, 8 Elements: 1 ug/l each of Be, Ce, Fe, in, Li, Mg, Pb, U in 1% HNO₃
Catalog # 91181.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Matrix:
    1% HNO₃
  • Pack type:
    HDPE bottle
  • Pk:
    500 mL
  • Description:
    NexION Setup Solution (8 Elements)
  • Seq:
    04
  • Hazard class description:
    Corrosive
  • UN No.:
    3264
  • Storage temperature:
    Ambient
  • Elements and Concentration:
    1 ug/l each of Be, Ce, Fe, in, Li, Mg, Pb, U

Specifications

About this item

Certifications: ISO 17025/17034